Lanolin, anhydrous USP
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Lanolin, anhydrous USP
Catalog # ACRO413005000
Supplier: Thermo Fisher Scientific
CAS Number: 8006-54-0
Jump to:
Product Details & Documents
Synonyms: Wool fat
- CAS Number:8006-54-0
- MDL Number:MFCD00081740
- Merck Index:12,05371
- Melting Pt:38…40 °C
- Density:0,932 to 0.945 g/cm³ (15 °C)
- Flash Pt:209 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:232-348-6
Documents
Specifications
Specification Test Results
- Appearance:Yellow Wax
- Infrared spectrum:Conforms
- Melting point:38 to 44 °C
- Water:≤0.25 %
- Additional info:Water-soluble acids and alkalies: passes test
- Antioxidant:≤0.02 % (BHT)
- Residue after ignition:≤0.1 %
- Acidity:≤2.0 ml (ml 0.1 N NaOH)
- Alkalinity:Passes test
- Chloride (Cl):≤0.035 %
- Iodine value:18 to 36
- Ammonia (NH3):Passes test
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



