Orders LinkContactUsLinkComponent
Funnel holders
Funnel holders
Funnel holder for a funnel, 1 funnel
Catalog # 241-0343
Supplier:   Brand
Funnel holders
Catalog # 241-0343
Supplier:  Brand
Funnel holders
Funnel holders
Funnel holder for a funnel, 1 funnel
Price
74,50 €
Log in for availability
1 * 5 ST
Product Details & Documents
PP, white.
To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8 to 14 mm with a locking nut.
Specifications
  • For:
    1 funnel
  • Description:
    Funnel holder for a funnel