Separating funnels
1000 ml, 53 mm
Catalog # BRND140007
Supplier: Brand
Wechseln zu:
Product Details & Documents
PP, high clarity, PTFE stopcock, PP screw cap.
- Leakproof closure and stopcock
- Excellent chemical resistance
- Autoclavable except for stopcock assembly
- PTFE stopcock requires no lubrication
High clarity for observation of contents.
Spezifikationen:
- Capacity:1000 ml
- Ø cap:53 mm



