Order Entry
Germany
Orders LinkContactUsLinkComponent
Separating funnels

Separating funnels

500 ml, 43 mm
Catalog # BRND140006
Supplier:   Brand
Separating funnels
Catalog # BRND140006
Supplier:  Brand
Separating funnels
Separating funnels

500 ml, 43 mm

Price
182,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, high clarity, PTFE stopcock, PP screw cap.

  • Leakproof closure and stopcock
  • Excellent chemical resistance
  • Autoclavable except for stopcock assembly
  • PTFE stopcock requires no lubrication

High clarity for observation of contents.

Specifications
  • Capacity:
    500 ml
  • Ø cap:
    43 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.