Order Entry
Germany
Orders LinkContactUsLinkComponent
Separating funnels

Separating funnels

250 ml, 33 mm
Catalog # BRND140005
Supplier:   Brand
Separating funnels
Catalog # BRND140005
Supplier:  Brand
Separating funnels
Separating funnels

250 ml, 33 mm

Wechseln zu:
Price
167,65 €
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents

PP, high clarity, PTFE stopcock, PP screw cap.

  • Leakproof closure and stopcock
  • Excellent chemical resistance
  • Autoclavable except for stopcock assembly
  • PTFE stopcock requires no lubrication

High clarity for observation of contents.

Spezifikationen:
  • Capacity:
    250 ml
  • Ø cap:
    33 mm