Separating funnels
125 ml, 28 mm
Catalog # BRND140004
Supplier: Brand
Jump to:
Product Details & Documents
PP, high clarity, PTFE stopcock, PP screw cap.
- Leakproof closure and stopcock
- Excellent chemical resistance
- Autoclavable except for stopcock assembly
- PTFE stopcock requires no lubrication
High clarity for observation of contents.
Specifications
- Capacity:125 ml
- Ø cap:28 mm



