Order Entry
Germany
Orders LinkContactUsLinkComponent
Ladle scoops

Ladle scoops

1000 ml, 350 mm, 160 mm
Catalog # 231-1105
Supplier:   BOCHEM
Ladle scoops
Catalog # 231-1105
Supplier:  BOCHEM
Ladle scoops
Ladle scoops

1000 ml, 350 mm, 160 mm

Price
77,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic and electropolished.

  • Complete with flat handle
  • Corrosion resistant
  • Excellent thermal resistance
Specifications
  • Ø:
    160 mm
  • Capacity:
    1000 ml
  • Length:
    350 mm