Ladle scoops
200 ml, 315 mm, 90 mm
Catalog # 231-1102
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic and electropolished.
- Complete with flat handle
- Corrosion resistant
- Excellent thermal resistance
Specifications
- Ø:90 mm
- Capacity:200 ml
- Length:315 mm



