Order Entry
Germany
Orders LinkContactUsLinkComponent
Ladle scoops

Ladle scoops

90 ml, 185 mm, 70 mm
Catalog # 231-1101
Supplier:   BOCHEM
Ladle scoops
Catalog # 231-1101
Supplier:  BOCHEM
Ladle scoops
Ladle scoops

90 ml, 185 mm, 70 mm

Price
15,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic and electropolished.

  • Complete with flat handle
  • Corrosion resistant
  • Excellent thermal resistance
Specifications
  • Ø:
    70 mm
  • Capacity:
    90 ml
  • Length:
    185 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.