Order Entry
Germany
Orders LinkContactUsLinkComponent
Clamps

Clamps

Stainless steel 18/10, 80 mm, 180 mm
Catalog # BOCH5409
Supplier:   BOCHEM
Clamps
Catalog # BOCH5409
Supplier:  BOCHEM
Clamps
Clamps

Stainless steel 18/10, 80 mm, 180 mm

Price
35,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Retort clamps made of stainless steel.

  • Jaws are covered with a layer of cork to protect clamped items
  • Angled jaws

Certifications: DIN 12894

Specifications
  • Spanning width:
    80 mm
  • Length:
    180 mm
  • Material:
    Stainless steel 18/10
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.