Order Entry
Germany
Orders LinkContactUsLinkComponent
Weighing scoops

Weighing scoops

105 mm, 32 mm
Catalog # 611-9712
Supplier:   BOCHEM
Weighing scoops
Catalog # 611-9712
Supplier:  BOCHEM
Weighing scoops
Weighing scoops

105 mm, 32 mm

Price
10,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

Specifications
  • Width:
    32 mm
  • Length:
    105 mm