Ethoxytrimethylsilane 95%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Ethoxytrimethylsilane 95%
Catalog # ACRO313820250
Supplier: Thermo Fisher Scientific
CAS Number: 1825-62-3
Jump to:
Product Details & Documents
Synonyms: Trimethyl(ethoxy)silane
- CAS Number:1825-62-3
- Formula:C₅H₁₄OSi
- MDL Number:MFCD00009069
- Molecular Weight:118.25 g/mol
- Boiling Pt:75…76 °C (760 mmHg)
- Melting Pt:–83 °C
- Density:0.75 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:217-370-6
- UN:1993
- ADR:3,II
Documents
ACRO31382
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3730 to 1.3750 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



