Ethoxytrimethylsilane 95%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Ethoxytrimethylsilane 95%
Catalog # ACRO313820050
Supplier: Thermo Fisher Scientific
CAS Number: 1825-62-3
Jump to:
Product Details & Documents
Synonyms: Trimethyl(ethoxy)silane
- CAS Number:1825-62-3
- Formula:C₅H₁₄OSi
- MDL Number:MFCD00009069
- Molecular Weight:118.25 g/mol
- Boiling Pt:75…76 °C (760 mmHg)
- Melting Pt:–83 °C
- Density:0.75 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:217-370-6
- UN:1993
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3730 to 1.3750 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



