Orders LinkContactUsLinkComponent
Manikins
Manikins
Susie Simon Patient Care Simul.+Ostomy
Catalog # BINH1005785
Marketsource
Supplier:   3B Scientific
Manikins
Catalog # BINH1005785
Supplier:  3B Scientific
Marketsource
Manikins
Manikins
Susie Simon Patient Care Simul.+Ostomy
Price
3 710,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This full-size manikin allows the training of BLS rescue techniques and CPR. The resiliency and weight of the body is very realistic.
  • Several models
Specifications
  • Description:
    Susie Simon Patient Care Simul.+Ostomy