Funnels with short stem
150 mm, 16 mm, 240 - 270 mm, 150 mm
221-0007
PYREX®, borosilicate glass
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls.
- Funnel O-Ø:150 mm
- Stem O-Ø:16 mm
- Stem length:150 mm
- For filter Ø:240 - 270 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



