Order Entry
Germany
Orders LinkContactUsLinkComponent
Polysorbate 80

Polysorbate 80

Glass bottle, 1 L
Catalog # ACRO278630010
Polysorbate 80
Catalog # ACRO278630010
CAS Number:  9005-65-6
Polysorbate 80
Polysorbate 80

Glass bottle, 1 L

CAS Number:  9005-65-6
Price
111,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:-21 °C
  • Density:1.08 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to amber Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4700 to 1.4760 (20°C, 589 nm)
  • Water:
    ≤3 %
  • Acid value:
    ≤2 mg KOH/g
  • Hydroxyl value:
    65 to 80 mg KOH/g
  • Saponification value:
    45 to 55 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications