Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution pH 10, carbonate buffer

Buffer solution pH 10, carbonate buffer

10.00, ±0,02, Carbonate, Colourless, Plastic bottle, 1 L
Catalog # ACRO258600010
Buffer solution pH 10, carbonate buffer
Catalog # ACRO258600010
CAS Number:  
Buffer solution pH 10, carbonate buffer
Buffer solution pH 10, carbonate buffer

10.00, ±0,02, Carbonate, Colourless, Plastic bottle, 1 L

CAS Number:  
Price
58,60 €
Log in for availability
1 * 1 L
Product Details & Documents

CAS No.: 1332-57-6

MDL No.: MFCD00036143
Appearance: Clear colourless solution
Storage Temperature: Ambient

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Carbonate
  • Pk:
    1 L
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    10.00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.