Order Entry
Germany
Orders LinkContactUsLinkComponent
Polysorbate 20

Polysorbate 20

Glass bottle, 1 L
Catalog # ACRO233360010
Polysorbate 20
Catalog # ACRO233360010
CAS Number:  9005-64-5
Polysorbate 20
Polysorbate 20

Glass bottle, 1 L

CAS Number:  9005-64-5
Price
72,80 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C (760 mmHg)
  • Density:1.1 g/cm³
  • Flash Pt:>150 °C
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear to slightly hazy yellow to brown-yellow Viscous liquid (May develop some turbidity or precipitate)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4680 to 1.4720 (20°C, 589 nm)
  • Water:
    ≤3 %
  • Dioxane:
    ≤10 ppm
  • Acid value:
    ≤2 mg KOH/g
  • Hydroxyl value:
    96 to 108 mg KOH/g
  • Saponification value:
    40 to 50 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.