Order Entry
Germany
Orders LinkContactUsLinkComponent
D-Cycloserine 98%

D-Cycloserine 98%

Glass bottle, 1 g
  ACRO228480010
 
D-Cycloserine 98%
  ACRO228480010
 :  68-41-7
D-Cycloserine 98%
D-Cycloserine 98%

Glass bottle, 1 g

 :  68-41-7
 
76,40 €
   
1 * 1 g
 
 :  D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
  •  :68-41-7
  •  :C₃H₆N₂O₂
  •  :MFCD00005353
  •  :102.09 g/mol
  •  :147 °C (dec.)
  •  :Refrigerator
  •  :200-688-4

Documents

 

 

  • Appearance:
    White to light yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (60°C, 3 hrs) (vacuum)
  • UV:
    at 285 nm A: ≤0.8
  • Condensation products:
    (0.04 % in 0.1 N NaOH)
  • Potency:
    ≥900 µg/mg
  • Residue after ignition:
    ≤0.5 %
  • pH:
    5.5 to 6.5 (10 % solution)
  • Specific optical rotation:
    +108° to +114° (20°C, 589 nm) (c=5, 2N NaOH)
  • Titration with NaOH:
    ≥97.5 %

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.