α-Cyclodextrine hydrate 98+%
Glass bottle, 25 g
: α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate
- :51211-51-9
- :C₃₆H₆₀O₃₀·xH₂O
- :MFCD00150804
- :972.86 g/mol (anhy)
- :278 °C (dec.)
- :Ambient
- :233-007-4
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤11 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.0 % (ELSD) on dry substance
- Specific optical rotation:+145° to +151° (20°C, 589 nm) (c=2, water) (on dry substance)
- Heavy metals (as Pb):≤15 ppm
- Pack type:Glass bottle
- Pk:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



