α-Cyclodextrine hydrate 98+%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
α-Cyclodextrine hydrate 98+%
Catalog # ACRO227290050
Supplier: Thermo Fisher Scientific
CAS Number: 51211-51-9
Jump to:
Product Details & Documents
Synonyms: α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate
- CAS Number:51211-51-9
- Formula:C₃₆H₆₀O₃₀·xH₂O
- MDL Number:MFCD00150804
- Molecular Weight:972.86 g/mol (anhy)
- Melting Pt:278 °C (dec.)
- Storage Temperature:Ambient
- EINECS:233-007-4
Documents
ACRO22729
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤11 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.0 % (ELSD) on dry substance
- Specific optical rotation:+145° to +151° (20°C, 589 nm) (c=2, water) (on dry substance)
- Heavy metals (as Pb):≤15 ppm
Specifications
- Pack type:Glass bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



