Order Entry
Germany
Orders LinkContactUsLinkComponent
Automatic burettes, Pellet pattern, lateral stopcock

Automatic burettes, Pellet pattern, lateral stopcock

50 ml, 0,1 ml, 500 mm, 0,075 ± ml
Catalog # HIRS341017560
Supplier:   HIRSCHMANN
Automatic burettes, Pellet pattern, lateral stopcock
Catalog # HIRS341017560
Supplier:  HIRSCHMANN
Automatic burettes, Pellet pattern, lateral stopcock
Automatic burettes, Pellet pattern, lateral stopcock

50 ml, 0,1 ml, 500 mm, 0,075 ± ml

Price
228,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Automatic burettes, Pellet pattern, class B, DURAN® with blue graduation.

  • Schellbach stripes for enhanced legibility
  • Lateral glass stopcock or PTFE valve stopcock

Certifications: Caliberated for DIN EN ISO 385.

Documents

Specifications
  • Capacity:
    50 ml
  • Division:
    0,1 ml
  • Tolerance:
    0,075 ± ml
  • Scale length:
    500 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.