Automatic burettes, Pellet pattern, lateral stopcock
25 ml, 0,05 ml, 500 mm, 0,045 ± ml
612-9804
Automatic burettes, Pellet pattern, class B, DURAN® with blue graduation.
- Schellbach stripes for enhanced legibility
- Lateral glass stopcock or PTFE valve stopcock
Certifications: Caliberated for DIN EN ISO 385.
- Capacity:25 ml
- Division:0,05 ml
- Tolerance:0,045 ± ml
- Scale length:500 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



