Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
Capillary inlet trap assembly
Catalog # HEWLG1544-80550
Supplier:   Agilent
Inlet liners
Catalog # HEWLG1544-80550
Supplier:  Agilent
Inlet liners
Inlet liners
Capillary inlet trap assembly
Price
374,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Specifications
  • Description:
    Capillary inlet trap assembly