Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
Catalog # HEWLG1328-87600
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWLG1328-87600
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
Price
111,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
  • Seq:
    28