Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, green, stainless steel, 380 mm, I.Ø 0,17 mm, to connect column to DAD
Catalog # HEWLG1315-87311
Supplier:   Agilent
Capillaries for LC systems
Catalog # HEWLG1315-87311
Supplier:  Agilent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, pre-swaged, green, stainless steel, 380 mm, I.Ø 0,17 mm, to connect column to DAD
Price
148,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
  • Description:
    Capillaries, pre-swaged, green, stainless steel, 380 mm, I.Ø 0,17 mm, to connect column to DAD
  • Seq:
    49