Capillaries for LC systems
Capillaries, 1 end pre-swaged, green, stainless steel, 180 mm, I.Ø 0,17 mm, to connect autosampler to column
Catalog # HEWLG1313-87305
Supplier: Agilent
Jump to:
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifications
- Description:Capillaries, 1 end pre-swaged, green, stainless steel, 180 mm, I.Ø 0,17 mm, to connect autosampler to column
- Seq:05