Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Inlet liners

Graphite O-ring, for splitless liner, temperatures above 350 °C, 9,63 mm, 6,52 mm
  HEWL5180-4173
 :   Agilent
Inlet liners
  HEWL5180-4173
 :  Agilent
Inlet liners
Inlet liners

Graphite O-ring, for splitless liner, temperatures above 350 °C, 9,63 mm, 6,52 mm

 
95,00 €
   
1 * 10 ST
 

These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.

  • Consistent high level of inertness
 
  • Ext.Ø:
    9,63 mm
  • Ø int.:
    6,52 mm
  • Description:
    Graphite O-ring, for splitless liner, temperatures above 350 °C
  • Seq:
    54