Capillary columns, HP-1
on 7" cage, Capillary, HP-1, 1,50 µm, 0,53 mm, 15 m
Catalog # 558-0402
Supplier: Agilent
Jump to:
Product Details & Documents
Excellent general purpose column – "Industry Standard".
- Non-polar
- Wide range of applications
- Superior performance for low molecular weight alcohols (
- High temperature limit
- Bonded and cross-linked
- Solvent rinsable
100% Dimethylpolysiloxane
Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
Specifications
- Column Format:Capillary
- Film thickness:1,50 µm
- Ø int.:0,53 mm
- Description:on 7" cage
- Length:15 m
- Seq:65
- Phase:HP-1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



