Capillary columns, HP-1
on 7" cage, Capillary, HP-1, 0,17 µm, 0,32 mm, 50 m
Catalog # HEWL19091Z-015
Supplier: Agilent
Jump to:
Product Details & Documents
Excellent general purpose column – "Industry Standard".
- Non-polar
- Wide range of applications
- Superior performance for low molecular weight alcohols (
- High temperature limit
- Bonded and cross-linked
- Solvent rinsable
100% Dimethylpolysiloxane
Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
Specifications
- Column Format:Capillary
- Film thickness:0,17 µm
- Ø int.:0,32 mm
- Description:on 7" cage
- Length:50 m
- Seq:52
- Phase:HP-1



