Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides, Clear, 2, 1,5 mm
Catalog # 631-9489
Cavity slides
Catalog # 631-9489
Cavity slides
Cavity slides
Cavity slides, Clear, 2, 1,5 mm
Price
137,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Cavity microscopy slides.
  • Ground edges
  • Made from clear glass
Suitable for staining applications in microscopy.
Specifications
  • No. of wells:
    2
  • Colour:
    Clear
  • Description:
    Cavity slides
  • Seq:
    02
  • Thickness:
    1,5 mm