Order Entry
Germany
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Straight stirring rod, Steel, 2 mm, 125 mm
Catalog # HAMMHWN070-12
Supplier:   HAMMACHER KARL
Stirring rods
Catalog # HAMMHWN070-12
Supplier:  HAMMACHER KARL
Stirring rods
Stirring rods

Straight stirring rod, Steel, 2 mm, 125 mm

Price
10,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stirring rods, made from WIRONIT steel. Choice of rods with straight or angled end.

  • Rust resistant
  • Anti-magnetic
Specifications
  • Ø:
    2 mm
  • Description:
    Straight stirring rod
  • Length:
    125 mm
  • Material:
    Steel
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.