Orders LinkContactUsLinkComponent
Dissecting scissors, type Joseph
Dissecting scissors, type Joseph
Joseph, curved, sharp/sharp, 140 mm
Catalog # HAMML-HSB029-15
Supplier:   HAMMACHER KARL
Dissecting scissors, type Joseph
Catalog # HAMML-HSB029-15
Supplier:  HAMMACHER KARL
Dissecting scissors, type Joseph
Dissecting scissors, type Joseph
Joseph, curved, sharp/sharp, 140 mm
Price
94,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dissecting scissors made of stainless steel.
Documents
Specifications
  • Type:
    Joseph, curved, sharp/sharp
  • Length:
    140 mm
  • Seq:
    02