Staining tiles
6, Ø×H: 36×11 mm, 80×115×12 mm
Catalog # 631-9201
Supplier: MORGAN ADVANCED MATERIALS HALD
Jump to:
Product Details & Documents
Glazed porcelain. These large tiles with cavities/wells are suitable for staining applications.
Farbe: Weiß.
Specifications
- Anz. Wells:6
- B×T×H:80×115×12 mm
- Lochgröße:Ø×H: 36×11 mm



