Auftragserfassung
Germany
AuftragsverfolgungKontakt
 

Sulfate standard solution

Sulfate standard solution, 500 ml (NIST), 50 mg/l as SO42-, 500 mL
Catalog # HACH257849
Supplier:   Hach
Sulfate standard solution
Catalog # HACH257849
Supplier:  Hach
CAS Number:  
Sulfate standard solution
Sulfate standard solution

Sulfate standard solution, 500 ml (NIST), 50 mg/l as SO42-, 500 mL

CAS Number:  
Price
36,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solution for accuracy checks in sulfate determinations.

Specifications

Specifications

  • Konzentration:
    50 mg/l as SO<sub>4</sub><sup>2-</sup>
  • VE:
    500 mL
  • Bezeichnung:
    Sulfate standard solution, 500 ml (NIST)

Specifications