Order Entry
Germany
Orders LinkContactUsLinkComponent
Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution, 15 mg/l as NO₃-N (NIST), 100 mL
Catalog # HACH2415132
Supplier:   Hach
Nitrogen-nitrate standard solution
Catalog # HACH2415132
Supplier:  Hach
CAS Number:  
Nitrogen-nitrate standard solution
Nitrogen-nitrate standard solution

Nitrogen-nitrate standard solution, 15 mg/l as NO₃-N (NIST), 100 mL

CAS Number:  
Price
18,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.

Documents

Specifications

Specifications

  • Concentration:
    15 mg/l as NO₃-N (NIST)
  • Pk:
    100 mL
  • Description:
    Nitrogen-nitrate standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.