Order Entry
Germany
Orders LinkContactUsLinkComponent
Silica standard solution

Silica standard solution

Silica standard solution, 10 mg/l as SiO2 (NIST), 500 mL
Catalog # HACH140349
Supplier:   Hach
Silica standard solution
Catalog # HACH140349
Supplier:  Hach
CAS Number:  
Silica standard solution

Silica standard solution, 10 mg/l as SiO2 (NIST), 500 mL

CAS Number:  
Price
50,80 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solutions for accuracy checks in silica determinations.

Specifications

Specifications

  • Concentration:
    10 mg/l as SiO<sub>2 </sub> (NIST)
  • Pk:
    500 mL
  • Description:
    Silica standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.