Order Entry
Germany
Orders LinkContactUsLinkComponent
Silica standard solution

Silica standard solution

Silica standard solution, 1 mg/l as SiO2 (NIST), 500 mL
Catalog # 705-2000
Supplier:   Hach
Silica standard solution
Catalog # 705-2000
Supplier:  Hach
CAS Number:  
Silica standard solution

Silica standard solution, 1 mg/l as SiO2 (NIST), 500 mL

CAS Number:  
Price
45,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solutions for accuracy checks in silica determinations.

Specifications

Specifications

  • Concentration:
    1 mg/l as SiO<sub>2 </sub> (NIST)
  • Pk:
    500 mL
  • Description:
    Silica standard solution

Specifications