Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Saccharometers

10 - 25 %, 0,1, 4…26 °C
  641-0003
 :   GERING
Saccharometers
  641-0003
 :  GERING
Saccharometers
Saccharometers

10 - 25 %, 0,1, 4…26 °C

 
256,00 €
   
1 * 1 ST
 

These saccharometers with temperature correction scale are suitable for the official testing in breweries especially on wort.

 
  • Temperature Range:
    4…26 °C
  • Seq:
    05
  • Division:
    0,1
  • Range:
    10 - 25 %