Saccharometers
0 - 20 %, 0,1, 4…26 °C
Catalog # 641-0000
Supplier: GERING
Jump to:
Product Details & Documents
These saccharometers with temperature correction scale are suitable for the official testing in breweries especially on wort.
Specifications
- Temperature Range:4…26 °C
- Seq:02
- Division:0,1
- Range:0 - 20 %



