Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Brix saccharimeters

60 - 90 °Brix, 1,0, 300 mm
Catalog # 614-0084
Supplier:   GERING
Brix saccharimeters
Catalog # 614-0084
Supplier:  GERING
Brix saccharimeters
Brix saccharimeters

60 - 90 °Brix, 1,0, 300 mm

Price
131,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Brix saccharimeters with a reference temperature of 20 °C.

Specifications
  • Length:
    300 mm
  • Seq:
    22
  • Division:
    1,0
  • Range:
    60 - 90 °Brix
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.