Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Brix saccharimeters

0 - 30 °Brix, 1,0, 300 mm
  635-0320
 :   GERING
Brix saccharimeters
  635-0320
 :  GERING
Brix saccharimeters
Brix saccharimeters

0 - 30 °Brix, 1,0, 300 mm

 
121,00 €
   
1 * 1 ST
 

Brix saccharimeters with a reference temperature of 20 °C.

 
  • Length:
    300 mm
  • Seq:
    20
  • Division:
    1,0
  • Range:
    0 - 30 °Brix