Filtration capsules, AcroPak™
Product Details & Documents
A filter for every step in your process. From drug discovery and basic research to process development, Pall Laboratory's AcroPak capsules come in a variety of membrane types and volume sizes.
- 100% integrity tested (where noted)
- Endotoxin level: <0,25 EU/ml using Limulus Amebocyte Lysate test
- Bacterial retention: 0,2 μm lot samples retain 10⁷ cfu/cm² of B. diminuta per modified ASTM F838
- Mycoplasma reduction: 0,1 μm lot samples retain 10⁷ cfu/cm² A. Laidlawii per modified ASTM F838
- Biological safety: Passes United States Pharmacopeia (USP) biological reactivity test in vivo <88>
- Sterilisation by gamma irradiation
Primary applications include: Buffer media and reagent manufacturing, protein purification and cell culture and bioburden filtration.
Supor PES (polyethersulfone):
With high flow and capacity, broad fluid compatibility, low protein adsorption, active ingredients and stabilisers, Pall PES membranes are best suited for filtration of preparative fluids such as buffers, media and bioprocess intermediates.
Supor EKV Asymmetric PES:
PES filters with asymmetric membrane layers have the same benefits of Supor PES (above), but with additional debris holding. This makes them ideal for high turbidity, high bioburden solutions. All Supor EKV products are a 0,65 µm pre-filter and a 0,2 µm asymmetric membrane.
Fluorodyne II PVDF (hydrophilic polyvinylidene fluoride):
Low extractables, low product and excipient adsorption, simple validation for the sterilisation of a wide range of formulations make PVDF membranes ideal for final filtration.
Documents
Certificates
Search by Lot NumberBSE TSE PALL12991.pdf
BSE/TSE Certificates
CoC_PALL12467_IG1118 CoC_PALL12941_IF7039 coc_PALL12941_UD6185 coc_PALL12991_FM4818 coc_PALL12991_FM5022
Pall Laboratory AcroPak™ Filtration Capsules Pall Laboratory Simplifying Sample Preparation for Research Laboratories
Product Literature
PALL12069_TDS_EN PALL12994_TDS_EN
Datasheet
Specifications
- Connection:Stepped hose barb
- Filtration area:1000 cm²
- Pore size:0,8/0,2 μm
- Description:AcroPak™ 1000
- Seq:7



