Order Entry
Germany
Orders LinkContactUsLinkComponent
Colour Reference Solutions, Reag.Ph.Eur.

Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution GY (greenish yellow) for testing the colour intensity acc. to Ph. Eur. GY1-GY7 Reag. Ph. Eur., Glass bottle, 125 mL
Catalog # REGEEP705
Supplier:   REAGECON
Colour Reference Solutions, Reag.Ph.Eur.
Catalog # REGEEP705
Supplier:  REAGECON
CAS Number:  
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution GY (greenish yellow) for testing the colour intensity acc. to Ph. Eur. GY1-GY7 Reag. Ph. Eur., Glass bottle, 125 mL

CAS Number:  
Price
193,00 €
Log in for availability
1 * 125 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Description:
    Colour reference solution GY (greenish yellow) for testing the colour intensity acc. to Ph. Eur. GY1-GY7 Reag. Ph. Eur.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.