Order Entry
Germany
Orders LinkContactUsLinkComponent
Colour Reference Solutions, Reag.Ph.Eur.

Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution B (brown) for testing the colour intensity acc. to Ph. Eur. B1-B9 Reag. Ph. Eur., Glass bottle, 100 mL
Catalog # REGEEP703
Supplier:   REAGECON
Colour Reference Solutions, Reag.Ph.Eur.
Catalog # REGEEP703
Supplier:  REAGECON
CAS Number:  
Colour Reference Solutions, Reag.Ph.Eur.
Colour Reference Solutions, Reag.Ph.Eur.

Colour reference solution B (brown) for testing the colour intensity acc. to Ph. Eur. B1-B9 Reag. Ph. Eur., Glass bottle, 100 mL

CAS Number:  
Price
229,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Colour reference solution B (brown) for testing the colour intensity acc. to Ph. Eur. B1-B9 Reag. Ph. Eur.

Specifications