CONC. TO MAKE IRON SOLUTION 20PPM 1 * 100 mL
Catalog # REGE5001600C
Lieferant: REAGECON
Frequently Bought Together
VWR®, Nitrile Gloves
Lieferant: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Lieferant: Dr. Weigert
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Lieferant: Dr. Weigert
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Arsenic standard solution, for limit tests
Lieferant: REAGECON
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Lieferant: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Lieferant: VWR Chemicals
Ethanol, GPR RECTAPUR®
Lieferant: VWR Chemicals
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Lieferant: VWR Collection