Mechanical cell counters
Manual cell counter, 0 − 999, 5, Metal
Catalog # 720-0810
Supplier: DENOMINATOR
Jump to:
Product Details & Documents
Metal case mechanical cell counters that can count from 0 to 999.
- Anti-slip strips
- Reset button
- Totalizer
Specifications
- Range:0 − 999
- Description:Manual cell counter
- Material:Metal
- Array:5