Snap caps for sample bottles, WHEATON®
22 mm, 4/8/12 ml
Catalog # WHEA242612
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Natural low density polyethylene.
- All sizes have tabs for easy removal
- Good chemical resistance
- Good temperature resistance
For WHEATON snap cap sample bottles.
Specifications
- Verschlussgröße:22 mm
- Für Flaschen:4/8/12 ml