Orders LinkContactUsLinkComponent
Eclipse™ Standard Surface Pipette Tips
Eclipse™ Standard Surface Pipette Tips
Extended, Flextop, UltraFine, 1250 µl, +, 88,9 mm, Clear, Universal pipettes, 576/pack, 4608/case, 8 packs/case
Catalog # LBCN1045-965-008-9
New Product
Supplier:   Labcon
Eclipse™ Standard Surface Pipette Tips
Catalog # LBCN1045-965-008-9
Supplier:  Labcon
New Product
Eclipse™ Standard Surface Pipette Tips
Eclipse™ Standard Surface Pipette Tips
Extended, Flextop, UltraFine, 1250 µl, +, 88,9 mm, Clear, Universal pipettes, 576/pack, 4608/case, 8 packs/case
Price
Inquire For Price
Log in for availability
1 * 576 ST
Product Details & Documents
Labcon's Eclipse™ standard surface pipette tips are made of high-quality, 100% pure virgin medical grade polypropylene. The resin is pre-tested to ensure no extractables are present.
  • Uniquely engineered to fit most popular brands of pipettes
  • Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
  • Tip sizes range from 10 to 1250 µl
  • Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
  • Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
  • Made in USA
Labcon resins are pre-tested to ensure no extractables, PFAS, or other compounds are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs.
Certifications: PFAS-free, Endotoxin-free, ATP-free, RNase/Dnase-free, Low Trace Metals (<1 ppm).
Specifications
  • Packed:
    576/pack, 4608/case, 8 packs/case
  • Sterile:
    +
  • Volume:
    1250 µl
  • Length:
    88,9 mm
  • Tip style:
    Extended, Flextop, UltraFine
  • Tip colour:
    Clear
  • For use with:
    Universal pipettes