Orders LinkContactUsLinkComponent
Quick Sample Access
Quick Sample Access
Quick sample access, CB 260, CB 260-O₂, CBF 260, CBF 260-O₂
Catalog # 390-1982
New Product
Supplier:   Binder
Quick Sample Access
Catalog # 390-1982
Supplier:  Binder
New Product
Quick Sample Access
Quick Sample Access
Quick sample access, CB 260, CB 260-O₂, CBF 260, CBF 260-O₂
Price
555,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Quick sample access - vewing window (130×250 mm) in the inner door for quick access.
  • Lets you open only a small section of the incubator instead of the whole chamber, which greatly protects the internal atmosphere and the cells Design keeps temperature, CO₂, humidity, and (if used) O₂ much more stable during routine access, which improves cell health and experimental reproducibility Opening just one segment limits how much warm, humid, CO₂‑rich air escapes, so the incubator recovers to setpoints faster after each door opening
More stable conditions from fewer fluctuations in temperature and gases support more consistent growth of sensitive cells, especially in shared incubators that are opened many times per day. Because each opening affects a smaller volume, the incubator needs less CO₂ (and N₂ in hypoxic mode) to return to the desired atmosphere, which can cut gas use by 40:80% compared with a single large inner door. Lower gas consumption extends cylinder lifetime, reduces cost and workload for changing bottles, and helps meet sustainability goals.
Specifications
  • Description:
    Quick sample access
  • Seq:
    3
  • For:
    CB 260, CB 260-O₂, CBF 260, CBF 260-O₂